Filtr membránový Pragopor 4 pr. 50mm (100ks) 1 * 100 KS
PRAG4/50
Frequently Bought Together
VWR® Weighing Paper, Grade 531
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Glass Fibre Filters
: VWR Collection
Pipette tips, Finntip™
: Thermo Fisher Scientific
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Thymolphthalein
: Thermo Fisher Scientific