Sodium hydroxide
Plastic bottle for solids, 1 kg
MACH10006AP1G1000
: MACH
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,08627
- :1390 °C (1013 hPa)
- :323 °C
- :2.13 g/cm³ (20 °C)
- :Ambient
- :215-185-5
- :1823
- :8,II
- :01-2119457892-27
- Pack type:Plastic bottle for solids
- Pk:1 kg