Acetone
Glass bottle, 1 L
Catalog # LACH20001AT0M1000
Supplier: Lach-ner S.R.O.
Acetone
Catalog # LACH20001AT0M1000
Supplier: Lach-ner S.R.O.
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95.4 °C
- Density:0.792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:LACH20001AT0M1000
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



