Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Válec odměrný vysoký PP průsvitný lisovaná stupnice 1000ml 1 * 1 KS
Catalog # GLAS177.303.07
Supplier:
GLASSCO
Expand image
Válec odměrný vysoký PP průsvitný lisovaná stupnice 1000ml 1 * 1 KS
Catalog # GLAS177.303.07
Supplier:
GLASSCO
Frequently Bought Together
Ammonium heptamolybdate tetrahydrate, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
Bottles, wide neck, round, short type, HDPE, with screw cap
Supplier: LAMAPLAST
VWR®, Silicone Lab Mat
Supplier: VWR Collection