ContactUsLinkComponent
High temperature bath circulator wch-8, capacity: 8 liter, with digital fuzzy control & digital lcd- back light system, temp. range: ambient +5°c - 250°c, bath dimension: 180 x 320 x 155 mm, pump: 15 l/min., timer: 99 hr. / 59 min., incl. stainless a 1 * 1 items
Catalog # WITGDH.WCH00408
Supplier:  witeg Labortechnik
High temperature bath circulator wch-8, capacity: 8 liter, with digital fuzzy control & digital lcd- back light system, temp. range: ambient +5°c - 250°c, bath dimension: 180 x 320 x 155 mm, pump: 15 l/min., timer: 99 hr. / 59 min., incl. stainless a 1 * 1 items
Catalog # WITGDH.WCH00408
Supplier:  witeg Labortechnik
Frequently Bought Together