Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Cyclohexanone

Cyclohexanone

Glass bottle, 900 mL
Catalog # MACH30111
Flammable
Irritant
Supplier:   MACH
Cyclohexanone
Catalog # MACH30111
Supplier:  MACH
CAS Number:  108-94-1
Cyclohexanone
Cyclohexanone

Glass bottle, 900 mL

CAS Number:  108-94-1
Price
Inquire For Price
Log in for availability
1 * 900 mL
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    900 mL

Specifications