DURACAL BUFFER PH 7.00 CL, 500 ML 1 * 500 mL
Catalog # HAMI238952
Supplier: HAMILTON BONADUZ
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Bottle top vacuum filtration systems, Corning®
Supplier: Corning
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
DURACAL BUFFER PH 1.68 1 * 500 mL
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Dispenser tips, PD-Tips II
Supplier: Brand
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
VWR®, Tubing, Silicone
Supplier: VWR Collection
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
Mortars, without pestles
Supplier: MORGAN ADVANCED MATERIALS HALD
DURACAL BUFFER PH 4.01 CL, 500 ML 1 * 500 mL
Supplier: HAMILTON BONADUZ