DURACAL BUFFER PH 4.01 CL, 500 ML 1 * 500 mL
Catalog # HAMI238951
Supplier: HAMILTON BONADUZ
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Low Temperature PC Slide Box
Supplier: VWR Collection
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
Papír filtrační kvalit. KA1M pr.125 1 * 100 KS
Supplier: PAP═RNA PERèTEJN SPOL