ContactUsLinkComponent
Combination pH puncture electrode, no cable, S7 connector head, Diaphragm: 2 x Single Pore, Material: Peek shaft, Electrolyte: Poli solve polymeric electrolyte, Length: 35 mm, Diameter: 6 mm, Temperature range: 0 bis 60 °C 1 * 1 items
Catalog # HAMI242067
Supplier:  HAMILTON BONADUZ
Combination pH puncture electrode, no cable, S7 connector head, Diaphragm: 2 x Single Pore, Material: Peek shaft, Electrolyte: Poli solve polymeric electrolyte, Length: 35 mm, Diameter: 6 mm, Temperature range: 0 bis 60 °C 1 * 1 items
Catalog # HAMI242067
Supplier:  HAMILTON BONADUZ
Frequently Bought Together