ContactUsLinkComponent
recombinant/native protein, vta1 (human) recombinant protein, human vta1 (np_057569, 1 a.a. - 307 a.a. ) full-length recombinant pro tein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5849
Supplier:  Abnova
recombinant/native protein, vta1 (human) recombinant protein, human vta1 (np_057569, 1 a.a. - 307 a.a. ) full-length recombinant pro tein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5849
Supplier:  Abnova