ContactUsLinkComponent
recombinant/native protein, fabp12 (human) recombinant protein, human fabp12 (np_001098751, 1 a.a. - 140 a.a.) full-length recombina nt protein with his tag expressed in escherichia coli. 1 * 50 µG
Catalog # ABNOP5401
Supplier:  Abnova
recombinant/native protein, fabp12 (human) recombinant protein, human fabp12 (np_001098751, 1 a.a. - 140 a.a.) full-length recombina nt protein with his tag expressed in escherichia coli. 1 * 50 µG
Catalog # ABNOP5401
Supplier:  Abnova