ContactUsLinkComponent
recombinant protein, proteoliposome, clec12a (human) recombinant protein, human clec12a full-length orf (adr82900.1) recombinant pro tein without tag. 1 * 10 µG
Catalog # ABNOH00160364-G01
Supplier:  Abnova
recombinant protein, proteoliposome, clec12a (human) recombinant protein, human clec12a full-length orf (adr82900.1) recombinant pro tein without tag. 1 * 10 µG
Catalog # ABNOH00160364-G01
Supplier:  Abnova