ContactUsLinkComponent
recombinant protein, pdcd1lg2 (human) recombinant protein, purified pdcd1lg2 (aah74766.1 19 a.a. - 121 a.a.) human recombinant prote in with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00080380-H02
Supplier:  Abnova
recombinant protein, pdcd1lg2 (human) recombinant protein, purified pdcd1lg2 (aah74766.1 19 a.a. - 121 a.a.) human recombinant prote in with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00080380-H02
Supplier:  Abnova