ContactUsLinkComponent
recombinant protein, tnfsf18 (human) recombinant protein, purified tnfsf18 (aah69111.1 50 a.a. - 177 a.a.) human recombinant protein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00008995-H01
Supplier:  Abnova
recombinant protein, tnfsf18 (human) recombinant protein, purified tnfsf18 (aah69111.1 50 a.a. - 177 a.a.) human recombinant protein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00008995-H01
Supplier:  Abnova