ContactUsLinkComponent
recombinant protein, cd28 (human) recombinant protein, purified cd28 (np_006130.1, 18 a.a. - 152 a.a.) human recombinant protein wit h his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00000940-H01
Supplier:  Abnova
recombinant protein, cd28 (human) recombinant protein, purified cd28 (np_006130.1, 18 a.a. - 152 a.a.) human recombinant protein wit h his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00000940-H01
Supplier:  Abnova