FENVALERATE D7 (ISOPROPYL D7) 1 * 1,1 mL
EHERXA13630010IO
Frequently Bought Together
pH buffers, ready to use
: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
Accessories for BagTools® Blender Bags
: INTERSCIENCE