DICLAZURIL-METHYL 1 * 10 mg
Catalog # EHERC12533200
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck