ContactUsLinkComponent
Cone, long tip, cone size 45/40, bore 31.0mm, shank outer diameter 42.0mm, shank length 90mm, wall thickness 2.30mm, useful for cons truction of stirrer guides, unbadged and free from ceramic markings, avoids the problem of the badge charring at high 1 * 50 items
Catalog # BIBBCBB45UB
Supplier:  SciLabware
Cone, long tip, cone size 45/40, bore 31.0mm, shank outer diameter 42.0mm, shank length 90mm, wall thickness 2.30mm, useful for cons truction of stirrer guides, unbadged and free from ceramic markings, avoids the problem of the badge charring at high 1 * 50 items
Catalog # BIBBCBB45UB
Supplier:  SciLabware
Frequently Bought Together