Hydrometer according to Casagrande, 350 mm 0.995-1.030:0.0005 g/cm , reference temp. 20°C 1 * 1 items
AMARH812020/WC
: Amarell
Hydrometer according to Casagrande, 350 mm 0.995-1.030:0.0005 g/cm , reference temp. 20°C 1 * 1 items
AMARH812020/WC
: Amarell
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection