ContactUsLinkComponent
MidiKros filter module TC 65cm 70kD mPES 1mm 1/2 TC X FLL sterile 1 * 1 items
Catalog # SPECT06-E070-10-S
MidiKros filter module TC 65cm 70kD mPES 1mm 1/2 TC X FLL sterile 1 * 1 items
Catalog # SPECT06-E070-10-S
Frequently Bought Together