ContactUsLinkComponent
MidiKros filter module TC 41,5cm 70kD mPES 1mm 1/2 TC X FLL dry 1 * 1 items
Catalog # SPECT04-E070-10-N
MidiKros filter module TC 41,5cm 70kD mPES 1mm 1/2 TC X FLL dry 1 * 1 items
Catalog # SPECT04-E070-10-N
Frequently Bought Together