Hydrochloric Acid 0.1N - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBC11-960
Supplier: REAGECON
Hydrochloric Acid 0.1N - 961.5ml of Conc. Dilutes 25L 1 * 6 L
Catalog # REGEDBC11-960
Supplier: REAGECON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Phthalic acid ≥99%
Supplier: Thermo Fisher Scientific