0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Fitting, PEEK HT, 1/16 one-piece hex-head short,natural, 10-32, 10'000 psi, 10/pkg, incl. ZNFT 1 * 10 items
JOURJR-5520N-10
:
VICI JOUR RESEARCH
Fitting, PEEK HT, 1/16 one-piece hex-head short,natural, 10-32, 10'000 psi, 10/pkg, incl. ZNFT 1 * 10 items
JOURJR-5520N-10
:
VICI JOUR RESEARCH
Frequently Bought Together
VWR® PH 20 pH Meter, Handheld
: VWR Collection
Accessories for titrators
: Mettler - Toledo
VWR® SP100 Automatic Straw Pipette, Fixed Volume
: VWR Collection
Potassium dihydrogen phosphate, EMSURE® ISO for analysis, Supelco®
: Merck
Accessories for Mettler balances
: Mettler - Toledo
Titration beakers
: Mettler - Toledo
pH/mV/°C meters, handheld, pH 3110 / 3310
: WTW
VWR®, Hand Tally Counter
: VWR Collection
Accessories for cell counter, EVE™
: NANOENTEK
VWR®, cell counting slides for automated cell counters
: VWR Collection
pH electrodes, SenTix® PLUS
: WTW
VWR® pHenomenal® Electrode Stand
: VWR Collection
ThermoMixer® C Thermomixer
: EPPENDORF
VWR® Facial Tissues
: VWR Collection
Accessories for OxiTop® and OxiTop® Control
: WTW
Orthophosphoric acid ≥85.0-88.0% NF, Multi-Compendial, J.T.Baker®
: Avantor
Plastic cuvette for LICO
: Hach