Entry port, 23 mm, standard position left centre top for unplus 1 * 1 items
MEMMUN450PLUSF1
: MEMMERT
Entry port, 23 mm, standard position left centre top for unplus 1 * 1 items
MEMMUN450PLUSF1
: MEMMERT
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals