ContactUsLinkComponent
Potential-free contact, 24 v/2 a, with socket to namur ne 28 for comination error message, e. g. supply failure, sensor fault, for u f 1 * 1 items
Catalog # MEMMUF260H6
Supplier:  MEMMERT
Potential-free contact, 24 v/2 a, with socket to namur ne 28 for comination error message, e. g. supply failure, sensor fault, for u f 1 * 1 items
Catalog # MEMMUF260H6
Supplier:  MEMMERT
Frequently Bought Together