11 COMPOSES 100 MG/L CHAQUE Al Cd Cr Cu Fe Ni Pb Zn P Mn Sn DANS HNO3 5% 1 * 100 mL
Catalog # CMPL75CA.K1.5N.L1
Supplier: CUSTOM MADE CHEMICALS LAB
11 COMPOSES 100 MG/L CHAQUE Al Cd Cr Cu Fe Ni Pb Zn P Mn Sn DANS HNO3 5% 1 * 100 mL
Catalog # CMPL75CA.K1.5N.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection