Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

9,00, ±0,02, Borate, Blue, 20 °C, 10 L
Catalog # 5659.9010
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Catalog # 5659.9010
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

9,00, ±0,02, Borate, Blue, 20 °C, 10 L

CAS Number:  
Price
7 920,00 Kč
Log in for availability
1 * 10 L
Product Details & Documents

The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).

  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.

Solutions are available in both clear and colour-coded varieties.

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Certifications: Each lot compared to NIST Standard Reference Buffer.

Specifications

Specifications

  • pH buffer type:
    Borate
  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,02
  • pH value:
    9,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.