Rivastigmine Related Compounds, Reference Standard
Rivastigmine Related Compound A, USP reference standard, Vial, 15 mg
Katalogové číslo 1604869.
Dodavatel: USP
Rivastigmine Related Compounds, Reference Standard
Katalogové číslo 1604869.
Dodavatel: USP
Číslo CAS:
Přejít na:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifikace
Specifikace
- Pack type:Vial
- Pk:15 mg
- Standards:USP reference standard
- Description:Rivastigmine Related Compound A
- Cat. No.:1604869.
- Formula:C₂₀H₁₈O₈
- Description:Rivastigmine Related Compound A
- Synonym(s):(+)-Di-(p-toluoyl)-D-tartaric acid
- CAS No.:32634-68-7



