Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Alclometasone Dipropionate, Reference Standards

Alclometasone Dipropionate, Reference Standards

Alclometasone Dipropionate Related Compound A, USP reference standard, 20 mg
Catalog # 1012746.
Supplier:   USP
Alclometasone Dipropionate, Reference Standards
Catalog # 1012746.
Supplier:  USP
CAS Number:  
Alclometasone Dipropionate, Reference Standards

Alclometasone Dipropionate Related Compound A, USP reference standard, 20 mg

CAS Number:  
Price
12 200,00 Kč
Log in for availability
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pk:
    20 mg
  • Standards:
    USP reference standard
  • Description:
    Alclometasone Dipropionate Related Compound A
  • Cat. No.:
    1012746.
  • Description:
    Alclometasone Dipropionate Related Compound A
  • Synonym(s):
    11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate

Specifications

Doporučení

Doporučení budou personalizována na základě vašich nákupních preferencí pouze v případě, že jste udělili souhlas aktivací možnosti "Vylepšit můj nákupní zážitek" na stránce "Osobní údaje - Nastavení ochrany osobních údajů".

V opačném případě obdržíte obecná doporučení.