ContactUsLinkComponent
Ng600a - 600ml/min ultra high purity nitrogen gas generator with built in compressors, psa, gc eu plug 1 * 1 items
Catalog # PEAK10-1070
Supplier:  PEAK SCIENTIFIC
undefined
Ng600a - 600ml/min ultra high purity nitrogen gas generator with built in compressors, psa, gc eu plug 1 * 1 items
Catalog # PEAK10-1070
Supplier:  PEAK SCIENTIFIC
Frequently Bought Together