ContactUsLinkComponent
15 COMPOSES N(NO3-) 210mg/l N(NH4+) 510mg/l P 625mg/l K 1205mg/l Ca 415mg/l Mg 120mg/l S 290mg/l B 10mg/l Cu 2.5mg/l Fe 2.5mg/l Mn 0.5mg/l Na 5mg/l Zn 0.5mg/l Cd 0.025mg/l Ni 0.05mg/l DANS HCL 3% 1 * 10.000 mL
Catalog # 89209.410
15 COMPOSES N(NO3-) 210mg/l N(NH4+) 510mg/l P 625mg/l K 1205mg/l Ca 415mg/l Mg 120mg/l S 290mg/l B 10mg/l Cu 2.5mg/l Fe 2.5mg/l Mn 0.5mg/l Na 5mg/l Zn 0.5mg/l Cd 0.025mg/l Ni 0.05mg/l DANS HCL 3% 1 * 10.000 mL
Catalog # 89209.410
Frequently Bought Together