ContactUsLinkComponent
Kit Template Generation System II (TGS II) generation of primer binding sites 20 reactions. Kit includes MuA Transposase (20l), 5x reaction buffer for MuA (100l), control target DNA (10l), Entranceposon (CamR-3) (20l), Entranceposon (KanR-3) (20 1 * 1 KIT
Catalog # F-702
Supplier:  FINNZYMES REAGENTS
Kit Template Generation System II (TGS II) generation of primer binding sites 20 reactions. Kit includes MuA Transposase (20l), 5x reaction buffer for MuA (100l), control target DNA (10l), Entranceposon (CamR-3) (20l), Entranceposon (KanR-3) (20 1 * 1 KIT
Catalog # F-702
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together