0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer pack PCR Phusion GC final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml of 50mM MgCl2 so lution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-519L
Supplier:
FINNZYMES REAGENTS
Expand image
Buffer pack PCR Phusion GC final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml of 50mM MgCl2 so lution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-519L
Supplier:
FINNZYMES REAGENTS
Frequently Bought Together
10 ml Precision FAST qPCR Master Mix for BioRad iCycler premixed with SYBRgreen (10x 1ml) 1 * 1.000 Tests
Supplier: PRIMERDESIGN
PFU DNA POLYMERASE 1 * 2.500 EU
Supplier: Abnova
GENELUTE BACTERIAL GENOMIC DNA KIT, 70 PREPS 1 * 1 items
Supplier: Merck