ContactUsLinkComponent
Kit DyNAmo cDNA synthesis 100x20µl (M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor), 2x RT buffer (includes dNTP mi x and 10 mM MgCl2), random hexamers (300 ng/µl), oligo(dT)15 primer (100 ng/µl)) 1 * 1 KIT
Catalog # F-470L
Supplier:  FINNZYMES REAGENTS
Kit DyNAmo cDNA synthesis 100x20µl (M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor), 2x RT buffer (includes dNTP mi x and 10 mM MgCl2), random hexamers (300 ng/µl), oligo(dT)15 primer (100 ng/µl)) 1 * 1 KIT
Catalog # F-470L
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together