ContactUsLinkComponent
Kit qPCR DyNAmo HS SYBR Green 1000x50µl reactions (2x Master Mix contains a hot start version of a modified Thermus brockianus DNA p olymerase, SYBR Green I dye, optimized PCR buffer, 5 mM MgCl2, dNTP mix including dUTP), vial of 50x ROX passive refe 1 * 1 KIT
Catalog # F-410XL
Supplier:  FINNZYMES REAGENTS
Kit qPCR DyNAmo HS SYBR Green 1000x50µl reactions (2x Master Mix contains a hot start version of a modified Thermus brockianus DNA p olymerase, SYBR Green I dye, optimized PCR buffer, 5 mM MgCl2, dNTP mix including dUTP), vial of 50x ROX passive refe 1 * 1 KIT
Catalog # F-410XL
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together