ContactUsLinkComponent
[FR] 37 COMPOSES 10 MG/L CHAQUE Ag Al As B Ba Be Bi Ca Cd Co Cr Cu Fe Ga Hg In K Li Mg Mn Mo Na Ni P Pb Sb Se Si Sn Sr Ta Ti Tl V W Zn Zr DANS HNO3 5% 1 * 250 mL
Catalog # 89175.230
[FR] 37 COMPOSES 10 MG/L CHAQUE Ag Al As B Ba Be Bi Ca Cd Co Cr Cu Fe Ga Hg In K Li Mg Mn Mo Na Ni P Pb Sb Se Si Sn Sr Ta Ti Tl V W Zn Zr DANS HNO3 5% 1 * 250 mL
Catalog # 89175.230
Frequently Bought Together