ContactUsLinkComponent
[FR]16 COMPOSES As 1000ug/ml Ba 300ug/ml Be 100ug/ml Cd 300ug/ml Cr 300ug/ml Co 300ug/ml Cu 300ug/ml Pb 1000ug/ml Mn 200ug/ml Ni 300ug/ml K 20000ug/ml Se 500ug/ml Ag 300ug/ml Tl 1000ug/ml V 300ug/ml Zn 300ug/ml DANS 1 * 50 mL
Catalog # 89162.150
[FR]16 COMPOSES As 1000ug/ml Ba 300ug/ml Be 100ug/ml Cd 300ug/ml Cr 300ug/ml Co 300ug/ml Cu 300ug/ml Pb 1000ug/ml Mn 200ug/ml Ni 300ug/ml K 20000ug/ml Se 500ug/ml Ag 300ug/ml Tl 1000ug/ml V 300ug/ml Zn 300ug/ml DANS 1 * 50 mL
Catalog # 89162.150
Frequently Bought Together