ContactUsLinkComponent
17 COMPOSES DANS HNO3 5% Aluminium (Al) 8mg/l Copper (Cu) 10mg/l Manganese (Mn) 10mg/l Cadmium (Cd) 10mg/l Arsenic (As) 40mg/l Iron (Fe) 40mg/l Barium (Ba) 100mg/l Lead (Pb) 100mg/l Antimony (Sb) 100mg/l Selenium (Se) 100mg/l Chro 1 * 100 mL
Catalog # 89144.180
17 COMPOSES DANS HNO3 5% Aluminium (Al) 8mg/l Copper (Cu) 10mg/l Manganese (Mn) 10mg/l Cadmium (Cd) 10mg/l Arsenic (As) 40mg/l Iron (Fe) 40mg/l Barium (Ba) 100mg/l Lead (Pb) 100mg/l Antimony (Sb) 100mg/l Selenium (Se) 100mg/l Chro 1 * 100 mL
Catalog # 89144.180
Frequently Bought Together