ContactUsLinkComponent
Burette 10:0.02ml CL.as with ns ptfeplug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 v ery good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422101
Supplier:  MARIENFELD
Burette 10:0.02ml CL.as with ns ptfeplug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 v ery good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422101
Supplier:  MARIENFELD
Frequently Bought Together