ContactUsLinkComponent
PRODUCT FOR RNA RELATE APPLICATION, SALMON SPERM DNA, SONICATED, SALMON SPERM DNA WAS SHARED BY SONICATION.DNA IS DISSOLVED IN DEION IZED DISTILLED WATER, WITH A FRAGMENT SIZE RANGE OF 300-700 BASE PAIRS. 1 * 5 mL
Catalog # ABNOR0014
Supplier:  Abnova
PRODUCT FOR RNA RELATE APPLICATION, SALMON SPERM DNA, SONICATED, SALMON SPERM DNA WAS SHARED BY SONICATION.DNA IS DISSOLVED IN DEION IZED DISTILLED WATER, WITH A FRAGMENT SIZE RANGE OF 300-700 BASE PAIRS. 1 * 5 mL
Catalog # ABNOR0014
Supplier:  Abnova