ContactUsLinkComponent
PRODUCT FOR RNA RELATE APPLICATION, SMALL RNA MARKER, SMALL RNAS INCLUDE A VARIETY OF NON-CODING RNAS, SUCH AS MIRNA, SIRNA, SNORNA AND SNRNA. RECENTLY SUCH A SMALL RNA IS INTENSIVELY STUDIED, BECAUSE THE SMALL RNA HAS BEEN FOUND TO CONTROL MANY BIOL 1 * 30 µl
Catalog # ABNOR0007
Supplier:  Abnova
PRODUCT FOR RNA RELATE APPLICATION, SMALL RNA MARKER, SMALL RNAS INCLUDE A VARIETY OF NON-CODING RNAS, SUCH AS MIRNA, SIRNA, SNORNA AND SNRNA. RECENTLY SUCH A SMALL RNA IS INTENSIVELY STUDIED, BECAUSE THE SMALL RNA HAS BEEN FOUND TO CONTROL MANY BIOL 1 * 30 µl
Catalog # ABNOR0007
Supplier:  Abnova