ContactUsLinkComponent
PRODUCT FOR RNA RELATE APPLICATION, RNA MARKER HIGH, THE RNA MARKER HIGH CONSISTS OF NINE SINGLE-STRANDED RNAS, 200, 500, 1,000, 1,5 00, 2,000, 3,000, 4,000, 5,000 AND 8,000 BASES, WHICH ARE SYNTHESIZED BY IN VITRO TRANSCRIPTION. THE RNA MARKER HIGH 1 * 50 µG
Catalog # ABNOR0003
Supplier:  Abnova
PRODUCT FOR RNA RELATE APPLICATION, RNA MARKER HIGH, THE RNA MARKER HIGH CONSISTS OF NINE SINGLE-STRANDED RNAS, 200, 500, 1,000, 1,5 00, 2,000, 3,000, 4,000, 5,000 AND 8,000 BASES, WHICH ARE SYNTHESIZED BY IN VITRO TRANSCRIPTION. THE RNA MARKER HIGH 1 * 50 µG
Catalog # ABNOR0003
Supplier:  Abnova
Frequently Bought Together