ContactUsLinkComponent
PRODUCT FOR RNA RELATE APPLICATION, RNA MARKER LOW, THE RNA MARKER LOW CONSISTS OF SEVEN SINGLE-STRANDED RNAS. THE 20-BASE AND 50-BA SE RNA ARE SYNTHESIZED BY CHEMICALLY (NOT PHOSPHORYLATED). THE 100, 200, 300, 400 AND 500 BASES ARE SYNTHESIZED BY IN 1 * 50 µG
Catalog # ABNOR0001
Supplier:  Abnova
PRODUCT FOR RNA RELATE APPLICATION, RNA MARKER LOW, THE RNA MARKER LOW CONSISTS OF SEVEN SINGLE-STRANDED RNAS. THE 20-BASE AND 50-BA SE RNA ARE SYNTHESIZED BY CHEMICALLY (NOT PHOSPHORYLATED). THE 100, 200, 300, 400 AND 500 BASES ARE SYNTHESIZED BY IN 1 * 50 µG
Catalog # ABNOR0001
Supplier:  Abnova