ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, CIB2 (HUMAN) RECOMBINANT PROTEIN, HUMAN CIB2 (NP_006374, 1 A.A. - 187 A.A.) FULL-LENGTH RECOMBINANT PROT EIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP3568
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, CIB2 (HUMAN) RECOMBINANT PROTEIN, HUMAN CIB2 (NP_006374, 1 A.A. - 187 A.A.) FULL-LENGTH RECOMBINANT PROT EIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP3568
Supplier:  Abnova