ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, FHIT (HUMAN) RECOMBINANT PROTEIN, HUMAN FHIT (NP_002003, 1 A.A. - 147 A.A.) FULL-LENGTH RECOMBINANT PROT EIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP3552
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, FHIT (HUMAN) RECOMBINANT PROTEIN, HUMAN FHIT (NP_002003, 1 A.A. - 147 A.A.) FULL-LENGTH RECOMBINANT PROT EIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP3552
Supplier:  Abnova