ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, TFAM (HUMAN) RECOMBINANT PROTEIN, HUMAN TFAM (NP_003192, 43 A.A. - 246 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 10 µG
  ABNOP3421
 :  Abnova
RECOMBINANT/NATIVE PROTEIN, TFAM (HUMAN) RECOMBINANT PROTEIN, HUMAN TFAM (NP_003192, 43 A.A. - 246 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 10 µG
  ABNOP3421
 :  Abnova
Frequently Bought Together