RECOMBINANT/NATIVE PROTEIN, RIBONUCLEASE A (BOVINE), LYOPHILIZED. PURIFIED. A PREPARATION EQUIVALENT TO CAT. # P5301. CONTAINS SOME AGGREGATES AS A RESULT OF LYOPHILIZATION BUT SHOWS NO DIFFERENCE IN SPECIFIC ACTIVITY. 1 * 25 mg
Catalog # ABNOP5301
Supplier: Abnova
Frequently Bought Together