ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, HEXOKINASE (YEAST), LYOPHILIZED. PHOSPHOHEXOSE ISOMERASE, 6-PHOSPHOGLUCONATE DEHYDROGENASE, ADENYLATE KI NASE AND CREATINE PHOSPHOKINASE CONTAMINANT ACTIVITIES ARE &LE 0.10%, 0.005%, 0.005% AND 0.005%, RESPECTIVELY. 1 * 2.500 EU
Catalog # ABNOP5230
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, HEXOKINASE (YEAST), LYOPHILIZED. PHOSPHOHEXOSE ISOMERASE, 6-PHOSPHOGLUCONATE DEHYDROGENASE, ADENYLATE KI NASE AND CREATINE PHOSPHOKINASE CONTAMINANT ACTIVITIES ARE &LE 0.10%, 0.005%, 0.005% AND 0.005%, RESPECTIVELY. 1 * 2.500 EU
Catalog # ABNOP5230
Supplier:  Abnova