ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, GP120 (89.6P) (HIV-1/CLADE B) RECOMBINANT PROTEIN, GP120 (89.6P) (HIV-1/CLADE B) (U89134, 34 A.A. - 518 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
  ABNOP5087
 :  Abnova
RECOMBINANT/NATIVE PROTEIN, GP120 (89.6P) (HIV-1/CLADE B) RECOMBINANT PROTEIN, GP120 (89.6P) (HIV-1/CLADE B) (U89134, 34 A.A. - 518 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
  ABNOP5087
 :  Abnova
Frequently Bought Together