ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, H5 (H5N1) (A/GOOSE/GUIYANG/337/2006) RECOMBINANT PROTEIN, H5 (H5N1) (A/GOOSE/GUIYANG/337/2006) (ABJ96698 , 17 A.A. - 530 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5069
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, H5 (H5N1) (A/GOOSE/GUIYANG/337/2006) RECOMBINANT PROTEIN, H5 (H5N1) (A/GOOSE/GUIYANG/337/2006) (ABJ96698 , 17 A.A. - 530 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5069
Supplier:  Abnova